siRNA Details
Gene Symbol: Spic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011461.3 | NP_035591.3 | 6 | 482 | ||
GenBank mRNA | AF098863.2 | - | 6 | 481 | ||
AK137522.1 | - | 6 | 485 | |||
AK156024.1 | - | 6 | 483 | |||
AK156892.1 | - | 6 | 483 | |||
AK157188.1 | - | 7 | 577 | |||
BC064038.1 | - | 5 | 443 | 3824951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
