siRNA Details
Gene Symbol: Spink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009258.5 | NP_033284.1 | 1, 2 | 143 | ||
GenBank mRNA | AK007841.1 | - | 1, 2 | 104 | ||
AK007985.1 | - | 1, 2 | 102 | |||
AV048854.1 | - | 1, 2 | 33 | |||
BC086887.1 | - | 1, 2 | 102 | 6775994 | ||
CA481615.1 | - | 1, 2 | 180 | |||
X06342.1 | - | 1, 2 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
