siRNA Details
Gene Symbol: Sprr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009264.2 | NP_033290.1 | 2 | 626 | ||
GenBank mRNA | AK144782.1 | - | 2 | 640 | ||
BC027534.1 | - | 2 | 626 | 1362548 | ||
X91824.1 | - | 2 | 619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
