siRNA Details
Gene Symbol: Sprr2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011468.4 | NP_035598.2 | 2 | 2636 | ||
GenBank mRNA | AK147183.1 | - | 2 | 2552 | ||
AK168529.1 | - | 2 | 2549 |
Gene Symbol: Sprr2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164787.1 | NP_001158259.1 | 2636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
