siRNA Details
Gene Symbol: Sprr2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011474.3 | NP_035604.1 | 2 | 591 | ||
GenBank mRNA | AJ005566.1 | - | 1 | 563 | ||
AY158992.1 | - | 2 | 591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
