siRNA Details
Gene Symbol: Sptlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011479.3 | NP_035609.1 | 6 | 822 | ||
GenBank mRNA | AK044917.1 | - | 6 | 823 | ||
AK057644.1 | - | 6 | 858 | |||
AK089019.1 | - | 6 | 816 | |||
AK159346.1 | - | 6 | 823 | |||
BC003227.1 | - | 6 | 855 | 3483158 | ||
U27455.1 | - | 6 | 790 | |||
X95642.1 | - | 6 | 793 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001037097, XM_008764823, BC107662.
Gene Ontology Categories:
