siRNA Details
Gene Symbol: Srm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009272.4 | NP_033298.1 | 6 | 933 | ||
GenBank mRNA | AK028359.1 | - | 6 | 944 | ||
AK051101.1 | - | 6 | 929 | |||
AK146395.1 | - | 6 | 890 | |||
BC005566.1 | - | 6 | 937 | 3966710 | ||
L19311.1 | - | 6 | 901 |
