siRNA Details
Gene Symbol: Ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166480.1 | NP_001159952.1 | 4 | 519 | ||
NM_009279.3 | NP_033305.1 | 4 | 401 | |||
GenBank mRNA | AK003044.1 | - | 4 | 381 | ||
AK007921.1 | - | 4 | 357 | |||
AK013723.1 | - | 4 | 363 | |||
AK142600.1 | - | 1 | 2957 | |||
AK160089.1 | - | 4 | 367 | |||
BC008172.1 | - | 4 | 322 | 3481948 | ||
BC012971.1 | - | 2 | 2058 | |||
BC019419.1 | - | 3 | 1852 | |||
BI989805.1 | - | 4 | 519 | |||
BQ747160.1 | - | 4 | 304 | |||
X90582.1 | - | 4 | 482 |
