siRNA Details
Gene Symbol: Stac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016853.2 | NP_058549.1 | 4 | 644 | ||
GenBank mRNA | AK134777.1 | - | 3 | 374 | ||
AK143440.1 | - | 4 | 677 | |||
BC107325.1 | - | 4 | 568 | 40054255 | ||
BC107326.1 | - | 4 | 568 | 40054256 | ||
D86639.1 | - | 4 | 644 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
