siRNA Details
Gene Symbol: Stat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205313.1 | NP_001192242.1 | 10, 11 | 1254 | ||
NM_001205314.1 | NP_001192243.1 | 10, 11 | 1290 | |||
NM_009283.4 | NP_033309.3 | 10, 11 | 1305 | |||
GenBank mRNA | AK018544.1 | - | 10, 11 | 1256 | ||
AK039458.1 | - | 10, 11 | 1306 | |||
AK046517.1 | - | 10, 11 | 1254 | |||
AK082706.1 | - | 10, 11 | 1287 | |||
AK084855.1 | - | 10, 11 | 1239 | |||
AK157263.1 | - | 10, 11 | 1154 | |||
AK159887.1 | - | 10, 11 | 1289 | |||
BC004808.1 | - | 10, 11 | 1241 | 3587831 | ||
U06924.1 | - | 8, 9 | 959 |
