siRNA Details
Gene Symbol: Stat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205313.1 | NP_001192242.1 | 13, 14 | 1433 | ||
NM_001205314.1 | NP_001192243.1 | 13, 14 | 1469 | |||
NM_009283.4 | NP_033309.3 | 13, 14 | 1484 | |||
GenBank mRNA | AK018544.1 | - | 13, 14 | 1435 | ||
AK039458.1 | - | 13, 14 | 1485 | |||
AK046517.1 | - | 13, 14 | 1433 | |||
AK082706.1 | - | 13, 14 | 1466 | |||
AK084855.1 | - | 13, 14 | 1418 | |||
AK157263.1 | - | 13, 14 | 1333 | |||
AK159887.1 | - | 13, 14 | 1468 | |||
BC004808.1 | - | 13, 14 | 1420 | 3587831 | ||
U06924.1 | - | 10, 11 | 1138 |
