siRNA Details
Gene Symbol: CXCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001565.3 | NP_001556.2 | 2 | 208 | ||
GenBank mRNA | AA321649.1 | - | 2 | 208 | ||
BC010954.1 | - | 2 | 173 | 4274617 | ||
X02530.1 | - | 2 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
