siRNA Details
Gene Symbol: Stat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011486.4 | NP_035616.1 | 3 | 474 | ||
NM_213659.2 | NP_998824.1 | 3 | 474 | |||
NM_213660.2 | NP_998825.1 | 3 | 474 | |||
GenBank mRNA | AK145486.1 | - | 3 | 330 | ||
AK153005.1 | - | 3 | 386 | |||
AK153472.1 | - | 3 | 386 | |||
AY299489.1 | - | 2 | 201 | |||
AY299490.1 | - | 2 | 201 | |||
BC003806.1 | - | 3 | 362 | 3593588 | ||
BC019168.1 | - | 3 | 378 | 4923137 | ||
L29278.1 | - | 3 | 453 | |||
U06922.1 | - | 3 | 263 | |||
U08378.1 | - | 3 | 341 | |||
U30709.1 | - | 3 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
