siRNA Details
Gene Symbol: Stat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011486.4 | NP_035616.1 | 3 | 470 | ||
NM_213659.2 | NP_998824.1 | 3 | 470 | |||
NM_213660.2 | NP_998825.1 | 3 | 470 | |||
GenBank mRNA | AK145486.1 | - | 3 | 326 | ||
AK153005.1 | - | 3 | 382 | |||
AK153472.1 | - | 3 | 382 | |||
AY299489.1 | - | 2 | 197 | |||
AY299490.1 | - | 2 | 197 | |||
BC003806.1 | - | 3 | 358 | 3593588 | ||
BC019168.1 | - | 3 | 374 | 4923137 | ||
L29278.1 | - | 3 | 449 | |||
U06922.1 | - | 3 | 259 | |||
U08378.1 | - | 3 | 337 | |||
U30709.1 | - | 3 | 363 |
