siRNA Details
Gene Symbol: Stat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY042906.1 | - | 11 | 1789 | ||
AY044901.1 | - | 11 | 1789 | |||
AY044902.1 | - | 11 | 1789 | |||
AY044903.1 | - | 11 | 1789 | |||
U21110.2 | - | 11 | 1789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
