siRNA Details
Gene Symbol: Stc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011491.3 | NP_035621.1 | 2, 3 | 503 | ||
GenBank mRNA | AF031035.1 | - | 2, 3 | 503 | ||
AF056244.1 | - | 2, 3 | 350 | |||
AK002527.1 | - | 2, 3 | 311 | |||
AK010318.1 | - | 2, 3 | 318 | |||
BC012206.1 | - | 2, 3 | 335 | 3665803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
