siRNA Details
Gene Symbol: Stfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082545.1 | NP_001076014.1 | 2 | 188 | ||
GenBank mRNA | BC138004.1 | - | 2 | 282 | 8861023 | |
BY780023.1 | - | 2 | 188 | |||
M92418.1 | - | 2 | 160 |
Gene Symbol: Stfa2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173869.3 | NP_776294.1 | 2 | 184 | ||
GenBank mRNA | AK050597.1 | - | 2 | 185 | ||
AY163161.1 | - | 2 | 160 | |||
BC117052.1 | - | 2 | 182 | 40126371 | ||
BC119354.1 | - | 2 | 182 | 8734103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
