siRNA Details
Gene Symbol: Stfa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025288.2 | NP_079564.1 | 3 | 281 | ||
GenBank mRNA | AK014349.1 | - | 3 | 284 | ||
AY169236.1 | - | 3 | 264 | |||
BC107224.1 | - | 3 | 275 | 40051891 | ||
BC107225.1 | - | 3 | 277 | 40051892 | ||
M92419.1 | - | 3 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
