siRNA Details
Gene Symbol: Stim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009287.4 | NP_033313.2 | 12 | 3301 | ||
GenBank mRNA | AK041944.1 | - | 12 | 3320 | ||
AK160523.1 | - | 1 | 1148 | |||
BC021644.1 | - | 12 | 3303 | 4191156 | ||
U47323.1 | - | 12 | 2903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
