siRNA Details
Gene Symbol: Aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291185.1 | NP_001278114.1 | 8 | 1089 | ||
NM_011497.4 | NP_035627.1 | 8 | 1167 | |||
GenBank mRNA | AF007817.1 | - | 8 | 1114 | ||
AK077513.1 | - | 8 | 1045 | |||
AK085861.1 | - | 8 | 1041 | |||
AK145968.1 | - | 8 | 1055 | |||
AK169363.1 | - | 8 | 1044 | |||
AK189850.1 | - | 1 | 16 | |||
AK213221.1 | - | 1 | 18 | |||
AY336976.1 | - | 7 | 991 | |||
BC005425.1 | - | 8 | 1034 | 3590706 | ||
BC014711.1 | - | 8 | 1103 | 3968835 | ||
U69106.1 | - | 8 | 1025 | |||
U80932.1 | - | 8 | 1034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
