siRNA Details
Gene Symbol: Bhlhe40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011498.4 | NP_035628.1 | 4, 5 | 671 | ||
GenBank mRNA | AF010305.1 | - | 4, 5 | 603 | ||
AF364051.1 | - | 4, 5 | 603 | |||
AK048242.1 | - | 4, 5 | 672 | |||
AK090032.1 | - | 4, 5 | 625 | |||
AK154935.1 | - | 4, 5 | 624 | |||
BC010720.1 | - | 4, 5 | 614 | 3707474 | ||
Y07836.1 | - | 4, 5 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
