siRNA Details
Gene Symbol: Stx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025307.1 | NP_001020478.1 | 5 | 999 | ||
NM_001286543.1 | NP_001273472.1 | 6 | 1045 | |||
NM_011502.3 | NP_035632.1 | 5 | 945 | |||
NM_152220.2 | NP_689344.1 | 5 | 999 | |||
GenBank mRNA | AK038415.1 | - | 5 | 506 | ||
AK133830.1 | - | 5 | 522 | |||
AK170105.1 | - | 5 | 408 | |||
AK171135.1 | - | 5 | 542 | |||
BC024844.1 | - | 5 | 407 | 5357204 | ||
BC056949.1 | - | 6 | 1045 | |||
D29797.1 | - | 5 | 420 | |||
D29798.1 | - | 5 | 418 | |||
D29799.1 | - | 5 | 433 | |||
D29800.1 | - | 6 | 377 |
