siRNA Details
Gene Symbol: Stx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009294.3 | NP_033320.1 | 8 | 893 | ||
GenBank mRNA | AK145909.1 | - | 8 | 912 | ||
AK161971.1 | - | 8 | 886 | |||
BC005791.1 | - | 8 | 733 | 3603665 | ||
BC011491.1 | - | 8 | 893 | 4016871 | ||
BC052023.1 | - | 8 | 829 | 5711022 | ||
U76832.1 | - | 8 | 662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
