siRNA Details
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011507.3 | NP_035637.2 | 4 | 458 | ||
GenBank mRNA | AF058956.1 | - | 3 | 300 | ||
AK042987.1 | - | 4 | 413 | |||
AK045176.1 | - | 4 | 411 | |||
AK156263.1 | - | 4 | 413 | |||
AK167136.1 | - | 4 | 413 | |||
AK167339.1 | - | 4 | 697 | |||
AK172561.1 | - | 4 | 413 | |||
AK172633.1 | - | 4 | 413 | |||
BC043312.1 | - | 2 | 153 | |||
BC054425.1 | - | 3 | 211 | |||
BC080781.1 | - | 4 | 413 | 30470296 | ||
BI985474.1 | - | 4 | 450 | |||
EU234009.1 | - | 4 | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
