siRNA Details
Gene Symbol: Supt4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009296.1 | NP_033322.1 | 2 | 118 | ||
GenBank mRNA | AK146235.1 | - | 2 | 133 | ||
AK167941.1 | - | 2 | 132 | |||
AK168229.1 | - | 2 | 133 | |||
BC024391.1 | - | 2 | 113 | 5011064 | ||
BC061174.1 | - | 2 | 119 | 6592996 | ||
BC087923.1 | - | 2 | 94 | 30308013 | ||
BC141092.1 | - | 2 | 156 | 9055704 | ||
U43154.1 | - | 2 | 83 |
