siRNA Details
Gene Symbol: Supt4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009296.1 | NP_033322.1 | 1, 2 | 99 | ||
GenBank mRNA | AK002990.1 | - | 1, 2 | 112 | ||
AK146235.1 | - | 1, 2 | 114 | |||
AK167941.1 | - | 1, 2 | 113 | |||
AK168229.1 | - | 1, 2 | 114 | |||
BC024391.1 | - | 1, 2 | 94 | 5011064 | ||
BC061174.1 | - | 1, 2 | 100 | 6592996 | ||
BC087923.1 | - | 1, 2 | 75 | 30308013 | ||
BC141092.1 | - | 1, 2 | 137 | 9055704 | ||
U43154.1 | - | 1, 2 | 64 |
Gene Symbol: Gm3258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011509.2 | NP_035639.1 | 1 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
