siRNA Details
Gene Symbol: Abcc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044720.1 | NP_001038185.1 | 31, 32 | 3947 | ||
NM_011511.2 | NP_035641.1 | 32, 33 | 3986 | |||
NM_021041.2 | NP_066378.1 | 32, 33 | 3986 | |||
NM_021042.2 | NP_066379.2 | 31, 32 | 3881 | |||
GenBank mRNA | AF003531.1 | - | 31, 32 | 3881 | ||
AK048665.1 | - | 1, 2 | 112 | |||
AK052291.1 | - | 6, 7 | 568 | |||
D86037.2 | - | 32, 33 | 3930 | |||
D86038.2 | - | 32, 33 | 3930 | |||
DQ991969.1 | - | 6, 7 | 601 | |||
DQ991970.1 | - | 6, 7 | 601 | |||
U97066.1 | - | 32, 33 | 3986 |
