siRNA Details
Gene Symbol: Med22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033908.1 | NP_001029080.1 | 2, 3 | 439 | ||
NM_011513.2 | NP_035643.1 | 2, 3 | 439 | |||
GenBank mRNA | AK144744.1 | - | 2, 3 | 418 | ||
AK161053.1 | - | 2, 3 | 380 | |||
AK166250.1 | - | 2, 3 | 385 | |||
AK166474.1 | - | 2, 3 | 412 | |||
AK192960.1 | - | 1, 2 | 27 | |||
AK204926.1 | - | 1, 2 | 27 | |||
AK212258.1 | - | 1, 2 | 96 | |||
BC018225.1 | - | 2, 3 | 429 | 3968506 | ||
BG805496.1 | - | 2, 3 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
