siRNA Details
Gene Symbol: Svs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009300.3 | NP_033326.1 | 2 | 276 | ||
GenBank mRNA | AK020349.1 | - | 2 | 277 | ||
BC115604.1 | - | 2 | 277 | 40102636 | ||
BC115605.1 | - | 2 | 277 | 40102637 | ||
M35732.1 | - | 2 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
