siRNA Details
Gene Symbol: Svs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013679.2 | NP_038707.2 | 2, 3 | 145 | ||
GenBank mRNA | AK020303.1 | - | 2, 3 | 147 | ||
BC119247.1 | - | 2, 3 | 149 | 8733996 | ||
BC119249.1 | - | 2, 3 | 149 | 8733998 | ||
L11393.1 | - | 2, 3 | 137 | |||
S65753.1 | - | 2, 3 | 137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
