siRNA Details
Gene Symbol: Swap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009302.3 | NP_033328.3 | 5 | 773 | ||
GenBank mRNA | AF053974.1 | - | 5 | 665 | ||
AK019882.1 | - | 5 | 717 | |||
AK045295.1 | - | 5 | 745 | |||
AK146773.1 | - | 5 | 765 | |||
AK172990.1 | - | 5 | 710 | |||
BC065136.1 | - | 5 | 715 | 30546814 |
