siRNA Details
Gene Symbol: Syn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111015.1 | NP_001104485.1 | 10 | 1395 | ||
NM_013681.3 | NP_038709.1 | 10 | 1395 | |||
GenBank mRNA | AF096867.1 | - | 10 | 1396 | ||
AK029181.1 | - | 8 | 737 | |||
AK043584.1 | - | 5 | 1607 | |||
BC053427.1 | - | 10 | 1316 | |||
BC066004.1 | - | 10 | 1214 | |||
BC085129.1 | - | 10 | 1214 | 5701861 | ||
BM951908.1 | - | 4 | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
