siRNA Details
Gene Symbol: Syn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013681.3 | NP_038709.1 | 11 | 1569 | ||
GenBank mRNA | AF096867.1 | - | 11 | 1570 | ||
AK029181.1 | - | 9 | 911 | |||
BM951908.1 | - | 5 | 443 |

More Information
Other Species Targeted

Gene Ontology Categories:
