siRNA Details
Gene Symbol: Syngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009303.2 | NP_033329.1 | 2 | 355 | ||
NM_207708.1 | NP_997591.1 | 2 | 355 | |||
GenBank mRNA | AJ002306.1 | - | 2 | 335 | ||
AK002972.1 | - | 2 | 337 | |||
AK010442.1 | - | 2 | 322 | |||
AK044007.1 | - | 2 | 346 | |||
AK153096.1 | - | 2 | 322 | |||
AK153187.1 | - | 2 | 322 | |||
AK156134.1 | - | 2 | 339 | |||
BC048172.1 | - | 2 | 312 | |||
BC138728.1 | - | 2 | 382 | 8861750 | ||
BC138729.1 | - | 2 | 393 | 8861751 |

More Information
Other Species Targeted

Gene Ontology Categories:
