siRNA Details
Gene Symbol: Syngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009304.2 | NP_033330.1 | 4 | 817 | ||
GenBank mRNA | AF151985.1 | - | 4 | 814 | ||
AF364049.1 | - | 4 | 797 | |||
AJ002307.1 | - | 4 | 783 | |||
AK150234.1 | - | 4 | 817 | |||
AK157343.1 | - | 4 | 790 | |||
AK170681.1 | - | 4 | 817 | |||
AK206414.1 | - | 1 | 59 | |||
AK207465.1 | - | 1 | 59 | |||
AK208216.1 | - | 1 | 59 | |||
AK209714.1 | - | 1 | 59 | |||
AK214806.1 | - | 1 | 59 | |||
AK215405.1 | - | 1 | 59 | |||
AK216027.1 | - | 1 | 165 | |||
AK218231.1 | - | 1 | 59 | |||
AK220017.1 | - | 1 | 59 | |||
AK220066.1 | - | 1 | 59 | |||
BC004829.1 | - | 4 | 810 | 2646657 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
