siRNA Details
Gene Symbol: Syt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252341.1 | NP_001239270.1 | 11 | 1708 | ||
NM_001252342.1 | NP_001239271.1 | 10 | 1619 | |||
NM_009306.3 | NP_033332.1 | 10 | 1628 | |||
GenBank mRNA | AK078790.1 | - | 11 | 1680 | ||
AK133578.1 | - | 10 | 1620 | |||
AK164153.1 | - | 10 | 1520 | |||
BC042519.1 | - | 10 | 1272 | 5365461 | ||
BC048187.1 | - | 10 | 1606 | |||
D37792.1 | - | 11 | 1568 | |||
FR827897.1 | - | 8 | 1109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
