siRNA Details
Gene Symbol: Syt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252341.1 | NP_001239270.1 | 6, 7 | 1020 | ||
NM_009306.3 | NP_033332.1 | 5, 6 | 940 | |||
GenBank mRNA | AK078790.1 | - | 6, 7 | 992 | ||
AK133578.1 | - | 5, 6 | 931 | |||
AK164153.1 | - | 5, 6 | 832 | |||
BC042519.1 | - | 5, 6 | 584 | 5365461 | ||
BC048187.1 | - | 5, 6 | 918 | |||
D37792.1 | - | 6, 7 | 880 | |||
FR827897.1 | - | 3, 4 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
