siRNA Details
Gene Symbol: Tac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009311.2 | NP_033337.1 | 2 | 144 | ||
GenBank mRNA | BC117081.1 | - | 2 | 119 | 40126400 | |
BC119122.1 | - | 2 | 119 | 8733871 | ||
BC144737.1 | - | 2 | 119 | 9053285 | ||
D17584.1 | - | 2 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
