siRNA Details
Gene Symbol: Tacr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009313.5 | NP_033339.2 | 3 | 1488 | ||
GenBank mRNA | AK038558.1 | - | 2 | 1145 | ||
BC075631.1 | - | 3 | 1488 | 30536972 | ||
BC098332.1 | - | 2 | 1395 | 30618488 | ||
L27826.1 | - | 2 | 386 | |||
L27828.1 | - | 2 | 386 | |||
X62934.1 | - | 2 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
