siRNA Details
Gene Symbol: Taf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020614.2 | NP_065639.2 | 4 | 302 | ||
GenBank mRNA | AK015111.1 | - | 4 | 269 | ||
AK131963.1 | - | 4 | 276 | |||
AK165580.1 | - | 4 | 282 | |||
AK170313.1 | - | 4 | 268 | |||
BC057167.1 | - | 4 | 289 | 4954753 | ||
BC086643.1 | - | 4 | 265 | |||
BC125027.1 | - | 4 | 251 | 40061781 | ||
BC125028.1 | - | 4 | 251 | 40061782 |

More Information
Other Species Targeted

Gene Ontology Categories:
