siRNA Details
Gene Symbol: Taf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020614.2 | NP_065639.2 | 15 | 1666 | ||
GenBank mRNA | AK131963.1 | - | 15 | 1640 | ||
AK165580.1 | - | 15 | 1646 | |||
AK170313.1 | - | 15 | 1632 | |||
BC057167.1 | - | 15 | 1653 | 4954753 | ||
BC069907.1 | - | 14 | 1523 | 4951630 | ||
BC086643.1 | - | 15 | 1627 | |||
BC094035.1 | - | 14 | 1518 | 5029813 | ||
BC125027.1 | - | 15 | 1615 | 40061781 | ||
BC125028.1 | - | 15 | 1615 | 40061782 | ||
CK619940.1 | - | 2 | 269 |

More Information
Other Species Targeted

Gene Ontology Categories:
