siRNA Details
Gene Symbol: Tagln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178598.2 | NP_848713.1 | 3 | 309 | ||
GenBank mRNA | AF149291.1 | - | 3 | 300 | ||
AF465519.1 | - | 3 | 246 | |||
AK151576.1 | - | 3 | 309 | |||
AK167395.1 | - | 3 | 294 | |||
AK167922.1 | - | 3 | 291 | |||
AK171088.1 | - | 3 | 277 | |||
AK172892.1 | - | 3 | 248 | |||
AK195747.1 | - | 2 | 72 | |||
BC009076.1 | - | 3 | 291 | 2654381 | ||
BC049861.1 | - | 3 | 508 | 4981433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
