siRNA Details
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130457.1 | NP_001123929.1 | 6 | 1274 | ||
NM_134011.2 | NP_598772.1 | 6 | 1274 | |||
GenBank mRNA | AK028966.1 | - | 6 | 1252 | ||
AK075897.1 | - | 3 | 548 | |||
AK129245.1 | - | 6 | 1229 | |||
AK144813.1 | - | 6 | 1273 | |||
AK166760.1 | - | 7 | 1182 | |||
AK168088.1 | - | 6 | 1272 | |||
BC016483.1 | - | 6 | 1230 | 3155271 | ||
BC031155.1 | - | 6 | 1245 | 4976690 | ||
BC031910.1 | - | 6 | 1244 | 5362943 | ||
U89434.1 | - | 3 | 399 |

More Information
Other Species Targeted

Gene Ontology Categories:
