siRNA Details
Gene Symbol: Tbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011538.2 | NP_035668.2 | 1, 2 | 140 | ||
GenBank mRNA | AY654733.1 | - | 1, 2 | 116 | ||
BC140951.1 | - | 1, 2 | 140 | 9055563 | ||
BF607954.1 | - | 1, 2 | 175 | |||
U57331.1 | - | 1, 2 | 140 |

More Information
Other Species Targeted

Gene Ontology Categories:
