siRNA Details
Gene Symbol: Tbxa2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277265.1 | NP_001264194.1 | 3 | 1460 | ||
NM_009325.4 | NP_033351.1 | 4 | 1614 | |||
GenBank mRNA | AK158477.1 | - | 3 | 1433 | ||
BC089611.1 | - | 4 | 1587 | 6762421 | ||
D10849.1 | - | 3 | 1339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
