siRNA Details
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159750.1 | NP_001153222.1 | 7 | 803 | ||
NM_001159751.1 | NP_001153223.1 | 7 | 699 | |||
NM_011541.4 | NP_035671.1 | 7 | 806 | |||
GenBank mRNA | AK131928.1 | - | 8 | 990 | ||
AK136035.1 | - | 7 | 699 | |||
AK143593.1 | - | 7 | 664 | |||
BC006022.1 | - | 7 | 681 | 3590733 | ||
BC061490.1 | - | 7 | 698 | 2582185 | ||
BC083127.1 | - | 7 | 669 | 6390364 | ||
M18209.1 | - | 6 | 602 | |||
M18210.1 | - | 7 | 660 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001025735, XM_008763546, XM_006251720, XM_006237816, BC099141, and others.
Gene Ontology Categories:
