siRNA Details
Gene Symbol: Tcf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009328.2 | NP_033354.2 | 2 | 822 | ||
GenBank mRNA | BC027533.1 | - | 2 | 822 | 5143248 | |
U18658.1 | - | 2 | 862 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
