siRNA Details
Gene Symbol: Tcf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011545.1 | NP_035675.1 | 1 | 646 | ||
GenBank mRNA | AB009453.1 | - | 1 | 643 | ||
AF029753.1 | - | 1 | 633 | |||
AF035717.1 | - | 1 | 646 | |||
AF036945.1 | - | 1 | 684 | |||
AF047418.1 | - | 1 | 619 | |||
AK011575.1 | - | 1 | 661 | |||
AK135423.1 | - | 1 | 693 | |||
AK157289.1 | - | 1 | 662 | |||
AK161795.1 | - | 1 | 679 | |||
BC053525.1 | - | 1 | 653 | 6529241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
