siRNA Details
Gene Symbol: Tfap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159696.1 | NP_001153168.1 | 4 | 778 | ||
NM_009335.2 | NP_033361.2 | 4 | 837 | |||
GenBank mRNA | AK050973.1 | - | 4 | 755 | ||
AK141211.1 | - | 4 | 739 | |||
AK145166.1 | - | 4 | 787 | |||
AK145603.1 | - | 4 | 732 | |||
BC003778.1 | - | 4 | 711 | 3592762 | ||
KJ463836.1 | - | 4 | 633 | |||
X94694.1 | - | 4 | 816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
