siRNA Details
Gene Symbol: Prdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011563.5 | NP_035693.3 | 2 | 209 | ||
GenBank mRNA | AK005225.1 | - | 2 | 237 | ||
AK008433.1 | - | 2 | 214 | |||
AK010653.1 | - | 2 | 184 | |||
AK011963.1 | - | 157 | ||||
AK014765.1 | - | 1 | 167 | |||
AK088280.1 | - | 2 | 228 | |||
BC002034.1 | - | 2 | 217 | 3485635 | ||
BC081454.1 | - | 2 | 128 | 6818396 | ||
BC086783.1 | - | 1 | 95 | 5700069 | ||
U20611.1 | - | 2 | 171 | |||
U51679.1 | - | 2 | 146 | |||
X82067.1 | - | 2 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
